Ammonium persulfate (APS) 98+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Peroxidisulphuric acid diammonium salt, Ammonium persulphate
40116-0050
40116-5000
AC40116-0020
AC40116-0100
AC40116-0250
AC40116-1000
TS40116-0050EA
266
USD
TS40116-0050
TS40116-5000
TS40116-0020
TS40116-0100
TS40116-0250
TS40116-1000
Ammonium persulfate (APS) 98+% ACS
Ammonium peroxodisulphate (APS)
Formula:
(NH₄)₂S₂O₈ MW: 228.2 g/mol Melting Pt: 120 °C (dec.) Density: 1.98 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003390 CAS Number: 7727-54-0 EINECS: 231-786-5 UN: 1444 ADR: 5.1,III Merck Index: 13,00545 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Residue after ignition | ≤0.05 % |
Titration with KMnO4 | ≥98.0 % |
Chloride and Chlorate (as Cl) | ≤0.001 % |
Iron (Fe) | ≤0.001 % |
Manganese (Mn) | ≤0.5 ppm |
Heavy metals (as Pb) | ≤0.005 % |
Titrable free acid | ≤0.04 meq/g |
Insoluble matter | ≤0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...