Ammonium persulfate (APS) 98%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Peroxidisulphuric acid diammonium salt, Ammonium persulphate

20153-1000 AC20153-0010 AC20153-0050
TS20153-1000EA 44 USD
TS20153-1000 TS20153-0010 TS20153-0050
Ammonium persulfate (APS) 98%, Extra Pure
Ammonium peroxodisulphate (APS)
Formula: (NH₄)₂S₂O₈
MW: 228.2 g/mol
Melting Pt: 120 °C
Density: 1.98 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003390
CAS Number: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Titration with KMnO4 ≥97.5 %
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR