Ammonium dihydrogen phosphate 99% for HPLC

Supplier: Thermo Scientific Chemicals

Synonyms: Ammonium dihydrogenphosphateAmmonium phosphate, monobasicAmmonium Phosphate Monobasic

44738-2500
TS44738-2500EA 257.93 USD
TS44738-2500
Ammonium dihydrogen phosphate 99% for HPLC
Ammonium dihydrogen phosphate
Formula: NH₄H₂PO₄
MW: 115.03 g/mol
Melting Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Water ≤1.0 %
Solubility (10 % in water) Clear colorless
UV (10 % in water)
UV at 260 nm A: ≤0.05
UV at 280 nm A: ≤0.05
pH 4.0 to 5.0 (10 % in water)
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR