di-Potassium hydrogen phosphate 98+% ACS

Supplier: Thermo Scientific Chemicals

42419-5000 AC42419-0025 AC42419-0050 AC42419-0100
TS42419-5000EA 136 USD
TS42419-5000 TS42419-0025 TS42419-0050 TS42419-0100
di-Potassium hydrogen phosphate 98+% ACS
di-Potassium hydrogen phosphate
Formula: K₂HPO₄
Melting Pt: 340 °C
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Loss on drying ≤1.0 % (2 g, 105°C)
pH 8.5 to 9.6 (5% soln. at 25°C)
Titration excess HCl ≥98.0 %
Chloride (Cl) ≤0.003 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Sodium (Na) ≤0.05 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.01 %
Nitrogen compounds (as N) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR