2-Mercaptoethanol 99%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
12547-0100
12547-0010
12547-0025
12547-2500
TS12547-0100EA
60
USD
TS12547-0100
TS12547-0010
TS12547-0025
TS12547-2500
2-Mercaptoethanol 99%, pure
2-Mercaptoethanol
Formula:
HSCH₂CH₂OH Boiling Pt: 157…158 °C (760 mmHg) Melting Pt: –100 °C Density: 1.11 g/cm³ Flash Pt: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004890 CAS Number: 60-24-2 EINECS: 200-464-6 UN: 2966 ADR: 6.1,II Merck Index: 13,05894 |
|
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4990 to 1.5020 (20°C, 589 nm) |
Water | ≤0.5 % |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...