Potassium hydroxide ca. 85%, flake, extra_pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

23255-0010 23255-0050 232550250
TS23255-0010EA 66 USD
TS23255-0010 TS23255-0050 TS232550250
Potassium hydroxide ca. 85%, flake, extra_pure
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 12,07806

Order Now

Specification Test Results

Appearance White Flakes
Titration with HCl ≥85 %
Chloride (Cl) ≤80 ppm
Sulfate (SO4) ≤20 ppm
Potassium carbonate ≤1 % (K2CO3)
Iron (Fe) ≤4 ppm
Sodium (Na) ≤0.9 % (as NaOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR