Potassium hydroxide ∼ 85%, pellets for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

13406-0010 13406-0025 134060100 134060250 134062500
TS13406-0010EA 92 USD
TS13406-0010 TS13406-0025 TS134060100 TS134060250 TS134062500
Potassium hydroxide ∼ 85%, pellets for analysis
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 12,07806

Order Now

Specification Test Results

Appearance White Pellets
Titration with HCl ≥85 %
Chloride (Cl) ≤70 ppm
Phosphate (PO4) ≤4 ppm
Sulfate (SO4) ≤10 ppm
Potassium carbonate ≤1 % (K2CO3)
Aluminium (Al) ≤10 ppm
Arsenic (As) ≤0.5 ppm
Calcium (Ca) ≤10 ppm
Iron (Fe) ≤5 ppm
Nickel (Ni) ≤3 ppm
Silver (Ag) ≤0.5 ppm
Sodium (Na) ≤0.4 %
Heavy metals (as Pb) ≤10 ppm
Silicon dioxide (SiO2) ≤5 ppm
Nitrogen compounds (as N) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR