Ammonium cerium(IV) nitrate, pure

Supplier: Thermo Scientific Chemicals
Danger

21236-0010 AC21236-0050
TS21236-0010EA 291 USD
TS21236-0010 TS21236-0050
Ammonium cerium(IV) nitrate, pure
Ammonium cerium (IV) nitrate
Formula: (NH₄)₂Ce(NO₃)₆
Melting Pt: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00151121
CAS Number: 16774-21-3
UN: 3085
ADR: 5.1,II

Order Now

Specification Test Results

Appearance Orange Crystalline powder or crystals
Loss on drying ≤2 %
Titration ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR