Diethanolamine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11392-0010
11392-0025
11392-0250
TS11392-0010EA
83
USD
TS11392-0010
TS11392-0025
TS11392-0250
Diethanolamine 99%
Diethanolamine
Formula:
C₄H₁₁NO₂ Boiling Pt: 268 °C (760 mmHg) Melting Pt: 27…30 °C Density: 1.0955 g/cm³ Flash Pt: 176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002843 CAS Number: 111-42-2 EINECS: 203-868-0 UN: 3259 ADR: 8,III REACH: 01-2119488930-28 Merck Index: 13,03134 |
Specification Test Results
Appearance | Clear viscous liquid or low melting solid |
Infrared spectrum | Conforms |
Refractive index | 1.4760 to 1.4790 (20°C, 589 nm) |
Water | ≤0.2 % (K.F.) |
GC | ≥98.5 % |
Color scale | ≤20 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...