Taurine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-Aminoethanesulphonic acid, 2-Aminoethanesulfonic acid
16654-5000
16654-0025
16654-0250
16654-1000
TS16654-5000EA
252
USD
TS16654-5000
TS16654-0025
TS16654-0250
TS16654-1000
Taurine 99%
Taurine
Formula:
C₂H₇NO₃S Melting Pt: 300 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00008197 CAS Number: 107-35-7 EINECS: 203-483-8 Merck Index: 13,09163 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Loss on drying | ≤0.2 % |
Sulfated ash | ≤0.1 % |
Titration with NaOH | ≥98.5 % |
Chloride (Cl) | ≤110 ppm |
Sulfate (SO4) | ≤140 ppm |
Ammonium (NH4) | ≤200 ppm |
Arsenic (As) | ≤2 ppm |
Heavy metals (as Pb) | ≤20 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...