Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8% ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Trizma(R) baseTRIS baseTHAM2-Amino-2-(hydroxymethyl)-1,3-propanediolTris(hydroxymethyl)methylamine2-Amino-2-(hydroxymethyl)propane-1,3-diol2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris)Tromethamine

42457-5000 42457-1000 42457-0025
TS42457-5000EA 128 USD
TS42457-5000 TS42457-1000 TS42457-0025
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8% ACS
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Formula: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Boiling Pt: 219…220 °C (10 mmHg)
Melting Pt: >168.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://us.vwr.com/stibo/web/std.lang.all/76/00/2297600.jpg

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Water ≤2 % (K.F.)
UV (40 % in water) 1 cm cell
UV at 290 nm A: ≤0.2
Titration with HCl 99.8 to 100.1 % (On dry basis)
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR