Glycolic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Hydroxy acid, 2-Hydroxyacetic acid

15451-0025 15451-0250 15451-1000 15451-5000
TS15451-0025EA 498 USD
TS15451-0025 TS15451-0250 TS15451-1000 TS15451-5000
Glycolic acid 99%
Glycolic acid
Formula: C₂H₄O₃
Boiling Pt: 100 °C (760 mmHg)
Melting Pt: 72…80 °C
Flash Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004312
CAS Number: 79-14-1
EINECS: 201-180-5
UN: 3261
ADR: 8,II
Merck Index: 13,04511

Order Now

Specification Test Results

Appearance White to off-white Adhering crystals
Infrared spectrum Conforms
Melting point 72 to 80 °C
Water ≤1 %
Titration with NaOH ≥98.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR