Urea 98%, pearls, Extra Pure
Supplier: Thermo Scientific Chemicals
19746-0050
19746-0010
19746-0100
TS19746-0050EA
207
USD
TS19746-0050
TS19746-0010
TS19746-0100
Urea 98%, pearls, Extra Pure
Urea
Formula:
NH₂CONH₂ Melting Pt: 131…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008022 CAS Number: 57-13-6 EINECS: 200-315-5 REACH: 01-2119463277-33 Merck Index: 13,09935 |
|
Specification Test Results
Appearance | White Pearls |
Infrared spectrum | Conforms |
Melting point | 131 to 135 °C |
Assay | ≥97.5 % (ex N) |
Water | ≤0.4 % |
Impurity | ≤1.2 % (Biuret) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...