Barium carbonate 99+% for analysis

Supplier: Thermo Scientific Chemicals
Warning

21749-0025 AC21749-5000
TS21749-0025EA 293 USD
TS21749-0025 TS21749-5000
Barium carbonate 99+% for analysis
Barium carbonate
Formula: BaCO₃
MW: 197.34 g/mol
Boiling Pt: 1450 °C
Melting Pt: 1350
Density: 4.43 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
Merck Index: 13,00972

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric ≥99.0 %
Chloride (Cl) ≤300 ppm
Sulfide (S) ≤10 ppm
Calcium (Ca) ≤1000 ppm
Iron (Fe) ≤50 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤2000 ppm
Strontium (Sr) ≤10000 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.015 % (in dilute HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR