Ninhydrin ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2-Dihydroxyindan-1,3-dionTrioxohydrindene monohydrateNinhydrin monohydrate2,2-Dihydroxy-1,3-indanedioneIndanetrione hydrate2,2-Dihydroxyindane-1,3-dione1,2,3-Indantrione monohydrate

41572-0010 41572-0100 41572-1000 41572-0250
TS41572-0010EA 1246 USD
TS41572-0010 TS41572-1000 TS41572-0100 TS41572-0250
Ninhydrin ACS
Ninhydrin
Formula: C₉H₆O₄
MW: 178.14 g/mol
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
Merck Index: 13,06582
https://us.vwr.com/stibo/web/std.lang.all/70/01/2297001.jpg

Order Now

Specification Test Results

Appearance White to light brown Crystals
Infrared spectrum Conforms
Melting point Passes test
Solubility Passes test
Identification Passes test
Sensitivity (to amino acids) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR