Propylene carbonate, anhydrous 99.5%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Carbonic acid propylene glycol ester4-Methyl-1,3-dioxolan-2-onePropylene glycol carbonate

429261000 42926-8000
TS42926-8000EA 136.16 USD
TS42926-8000 TS429261000
Propylene carbonate, anhydrous 99.5%, AcroSeal™
Propylene carbonate
Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 240…243 °C (760 mmHg)
Melting Pt: –49 °C
Density: 1.204 g/cm³
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00005385
CAS Number: 108-32-7

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR