Pyridine 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Azabenzene, Azine
41854-0250
41854-5000
41854-0025
418540010
TS41854-0250EA
2192
USD
TS41854-0250
TS41854-5000
TS41854-0025
TS418540010
Pyridine 99+% ACS
Pyridine
Formula:
C₅H₅N MW: 79.1 g/mol Boiling Pt: 115 Melting Pt: -44 Density: 0.978 g/cm³ Flash Pt: 17 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011732 CAS Number: 110-86-1 EINECS: 203-809-9 UN: 1282 ADR: 3 II Merck Index: 13,08060 |
|
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.1 % (K.F.) |
Solubility | (in water) Passes test |
GC | ≥99.0 % |
Residue after evaporation | ≤0.002 % |
Chloride (Cl) | ≤10 ppm |
Sulfate (SO4) | ≤10 ppm |
Copper (Cu) | ≤5 ppm |
Reducing substances | Passes test |
Ammonia (NH3) | ≤20 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...