Osmium tetraoxide 4% (w/w) in water
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Osmic acid anhydride
, Osmic acid
, Osmium (VIII) oxide
31901-0100
TS31901-0100EA
264
USD
TS31901-0100
Osmium tetraoxide 4% (w/w) in water
Osmium tetraoxide
Formula:
O₄Os MW: 254.23 g/mol Density: 1.04 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00011150 CAS Number: 20816-12-0 EINECS: 244-058-7 UN: 3287 ADR: 6.1,II |
Specification Test Results
Appearance | Clear yellow Solution |
Strength | 3.92 to 4.08 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...