Antimony(III) chloride ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Antimony trichloride
40137-0025
40137-5000
AC40137-0050
AC40137-1000
TS40137-0025EA
1475
USD
TS40137-0025
TS40137-5000
TS40137-0050
TS40137-1000
Antimony(III) chloride ACS
Antimony (III) chloride
Formula:
SbCl₃ Boiling Pt: 223 °C (760 mmHg) Melting Pt: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011212 CAS Number: 10025-91-9 EINECS: 233-047-2 UN: 1733 ADR: 8,II Merck Index: 13,00713 |
Specification Test Results
Appearance | Semitransparent colorless Adhering crystals (May have a pink hue) |
Titration Iodimetric | ≥99.0 % |
Sulfate (SO4) | ≤0.005 % |
Arsenic (As) | ≤0.02 % |
Calcium (Ca) | ≤0.005 % |
Copper (Cu) | ≤0.001 % |
Iron (Fe) | ≤0.002 % |
Lead (Pb) | ≤0.005 % |
Potassium (K) | ≤0.01 % |
Sodium (Na) | ≤0.02 % |
Insoluble matter | ≤0.05 % (in chloroform) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...