Magnesium chloride, pure
Supplier: Thermo Scientific Chemicals
AC22321-0025
22321-0250
AC22321-0010
AC22321-1000
TS22321-0250EA
480
USD
TS22321-0250
TS22321-1000
TS22321-0025
TS22321-0010
Magnesium chloride, pure
Magnesium chloride
Formula:
MgCl₂ MW: 95.21 g/mol Boiling Pt: 1412 °C (760 mmHg) Melting Pt: 708 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011106 CAS Number: 7786-30-3 EINECS: 232-094-6 |
Specification Test Results
Appearance | White Powder (May contain gray to black particles) |
Titration Complexometric | ≥98.0 % (on dry substance) |
Water | ≤3 % (K.F.) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...