Calcium chloride, anhydrous 96% for biochemistry

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

19273-5000 AC19273-1000
TS19273-5000EA 234 USD
TS19273-5000 TS19273-1000
Calcium chloride, anhydrous 96% for biochemistry
Calcium chloride
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
Merck Index: 14,01659

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Argentometric ≥95.0 %
Phosphate (PO4) ≤0.001 %
Sulfate (SO4) ≤0.02 %
Ammonium (NH4) ≤0.005 %
Arsenic (As) ≤0.0001 %
Copper (Cu) ≤0.0005 %
Iron (Fe) ≤0.0025 %
Nickel (Ni) ≤0.0005 %
Heavy metals (as Pb) ≤5 ppm
Acidity (HCl) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR