Sulfuric acid 96.00% (±0.15%) in water, pure

Supplier: Thermo Scientific Chemicals
Danger

30207-0010 30207-0025 302070011 302070026 302070100
TS30207-0010EA 87 USD
TS30207-0010 TS30207-0025 TS302070011 TS302070026 TS302070100
Sulfuric acid 96.00% (±0.15%) in water, pure
Sulfuric acid
Formula: H₂SO₄
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 10 °C
Density: 1.83 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Order Now

Specification Test Results

Appearance Clear colorless Viscous liquid
Titration with NaOH 95.85 to 96.15 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR