Sulfuric acid 96% in water, extra pure

Supplier: Thermo Scientific Chemicals
Danger

13361-0011 13361-0025 133610010 133610026 133610250
TS13361-0011EA 78 USD
TS13361-0011 TS13361-0025 TS133610010 TS133610026 TS133610250
Sulfuric acid 96% in water, extra pure
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.83 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II
Merck Index: 14,08974

Order Now

Specification Test Results

Appearance Clear liquid
Color scale ≤10 APHA
Titration with NaOH 95 to 98 %
Chloride (Cl) ≤0.5 ppm
Arsenic (As) ≤0.03 ppm
Iron (Fe) ≤0.5 ppm
Heavy metals (as Pb) ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR