Sodium sulfate, anhydrous 99+% for HPLC

Supplier: Thermo Scientific Chemicals

Synonyms: Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt

19410-2500 AC19410-0500
TS19410-2500EA 247 USD
TS19410-2500 TS19410-0500
Sodium sulfate, anhydrous 99+% for HPLC
Sodium sulphate
Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429
Melting Pt: 888
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Water ≤0.1 %
UV (10 % in water)
Acidity/alkalinity ≤0.0002 meq/g
Titration with HClO4 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR