Sodium peroxodisulfate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Disodium peroxodisulphate
, Peroxydisulphuric acid disodium salt
, Disodium peroxodisulfate
, Sodium peroxidisulfate
, Sodium peroxidisulphate
, Sodium persulphate
, Sodium persulfate
, Peroxydisulfuric acid disodium salt
20202-0010
20202-5000
AC20202-0050
TS20202-0010EA
104.26
USD
TS20202-0010
TS20202-0050
TS20202-5000
Sodium peroxodisulfate 98+%
Sodium peroxodisulphate
Formula:
Na₂S₂O₈ MW: 238.11 g/mol Melting Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003501 CAS Number: 7775-27-1 EINECS: 231-892-1 UN: 1505 ADR: 5.1,III Merck Index: 13,08729 |
Specification Test Results
Appearance | White to almost white Powder or crystals |
Titration Iodometric | 98.0 to 102.0 % |
Water | ≤0.15 % |
Iron (Fe) | ≤10 ppm |
Heavy metals (as Pb) | ≤20 ppm |
Total nitrogen | ≤0.2 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...