Sodium dihydrogen phosphate, anhydrous 99% for biochemistry
Supplier: Thermo Scientific Chemicals
Synonyms:
Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic, Sodium dihydrogen phosphate
44798-0010
44798-2500
TS44798-0010EA
165
USD
TS44798-0010
TS44798-2500
Sodium dihydrogen phosphate, anhydrous 99% for biochemistry
Sodium dihydrogen phosphate
Formula:
NaH₂PO₄ MW: 119.98 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003527 CAS Number: 7558-80-7 |
Specification Test Results
Appearance | White Crystalline powder and/or chunks |
Loss on drying | ≤1 % (130°C) |
Solubility | (1 M in water) Clear colorless |
UV | at 280 nm A: ≤0.2 |
pH | 4.0 to 6.0 (1 M in water) |
Titration with HClO4 | ≥98.5 % |
Heavy metals (as Pb) | ≤0.001 % |
Insoluble matter | ≤0.2 % (in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...