Cesium nitrate 99.99% (trace metals basis) for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nitric acid cesium salt

19207-2500 AC19207-0010 AC19207-0500
TS19207-2500EA 795 USD
TS19207-2500 TS19207-0010 TS19207-0500
Cesium nitrate 99.99% (trace metals basis) for analysis
Cesium nitrate
Formula: CsNO₃
MW: 194.91 g/mol
Boiling Pt: 737
Melting Pt: 414
Density: 3.68 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010963
CAS Number: 7789-18-6
EINECS: 232-146-8
UN: 1451
ADR: 5.1 III
Merck Index: 12,02057

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤30 ppm
Aluminium (Al) ≤0.2 ppm
Barium (Ba) ≤10 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.2 ppm
Iron (Fe) ≤0.2 ppm
Lithium (Li) ≤0.5 ppm
Magnesium (Mg) ≤0.5 ppm
Manganese (Mn) ≤1 ppm
Potassium (K) ≤1 ppm
Rubidium (Rb) ≤5 ppm
Sodium (Na) ≤1 ppm
Strontium (Sr) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR