Sodium periodate 99% for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium metaperiodate

19838-1000 19838-5000 AC19838-0025
TS19838-1000EA 152 USD
TS19838-1000 TS19838-5000 TS19838-0025
Sodium periodate 99% for analysis
Sodium periodate
Formula: NaIO₄
MW: 213.89 g/mol
Melting Pt: 300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 3085
ADR: 5.1,I
Merck Index: 13,08714

Order Now

Specification Test Results

Appearance White to off-white Crystals
Titration Iodometric ≥98.5 %
pH 3.4 to 4.5 (5 % at 20°C)
Chloride (Cl) ≤300 ppm
Iodide (I) ≤10 ppm
Nitrate (NO3) ≤2000 ppm
Sulfate (SO4) ≤200 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR