Sodium chloride 99.5% Biochemistry

Supplier: Thermo Scientific Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

44730-0050 44730-0010 44730-2500
TS44730-0050EA 252 USD
TS44730-0050 TS44730-0010 TS44730-2500
Sodium chloride 99.5% Biochemistry
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now

Specification Test Results

Appearance White Powder
Titration Argentometric ≥99.4 %
Solubility (0.1 M in water) Clear colorless
UV at 280 nm A: ≤0.01
Color scale ≤15 APHA (0.1 M in water)
pH 5.0 to 9.0
Heavy metals (as Pb) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR