Sodium chloride, extra_pure

Supplier: Thermo Scientific Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

19409-0250 AC19409-0010 AC19409-0050
TS19409-0250EA 373 USD
TS19409-0250 TS19409-0010 TS19409-0050
Sodium chloride, extra_pure
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Argentometric ≥99.0 %
pH 5.0 to 8.0 (5 % in water at 20°C)
Sulfate (SO4) ≤100 ppm
Barium (Ba) ≤50 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤50 ppm
Potassium (K) ≤100 ppm
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR