Sodium chloride 99.5% analysis_for

Supplier: Thermo Scientific Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

20779-0010 20779-0050 AC20779-0250
TS20779-0010EA 80 USD
TS20779-0010 TS20779-0050 TS20779-0250
Sodium chloride 99.5% analysis_for
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Argentometric ≥99.4 %
pH 5.0 to 8.0 (5 % at 20°C)
Bromide (Br) ≤50 ppm
Iodide (I) ≤10 ppm
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤20 ppm
Barium (Ba) ≤10 ppm
Calcium (Ca) ≤20 ppm
Iron (Fe) ≤3 ppm
Potassium (K) ≤100 ppm
Heavy metals (as Pb) ≤5 ppm
Total nitrogen (as N) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR