Sodium hydrogen carbonate 99.5% for biochemistry

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium bicarbonate

44710-0010 44710-2500
TS44710-2500EA 38.6 USD
TS44710-2500 TS44710-0010
Sodium hydrogen carbonate 99.5% for biochemistry
Sodium hydrogen carbonate
Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270 °C
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
https://us.vwr.com/stibo/web/std.lang.all/05/81/16640581.jpg

Order Now

Specification Test Results

Appearance White Crystalline powder
Solubility Clear colorless
UV at 280 nm A: ≤0.02
UV at 260 nm A: ≤0.03
UV (10 % in water)
Titration with HCl 99.4 to 100.6 %
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤150 ppm
Ammonium (NH4) ≤5 ppm
Calcium (Ca) ≤100 ppm
Copper (Cu) ≤2 ppm
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Zinc (Zn) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR