Potassium bromide 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

43695-0500 AC43695-0100
TS43695-0500EA 288 USD
TS43695-0500 TS43695-0100
Potassium bromide 99.999% (trace metals basis)
Potassium bromide
Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
Merck Index: 13,07701

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Argentometric 33 % (K) Typical
Total trace metal impurities ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR