Sodium borohydride 99%, caplets, VenPure™ AF, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
44849-5000
AC44849-0100
AC44849-1000
TS44849-5000EA
304
USD
TS44849-5000
TS44849-0100
TS44849-1000
Sodium borohydride 99%, caplets, VenPure™ AF, pure
Sodium borohydride
Formula:
NaBH₄ Melting Pt: 360 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003518 CAS Number: 16940-66-2 UN: 1426 ADR: 4.3,I |
Specification Test Results
Appearance | White Free flowing caplets |
Titration with Na2S2O3 | ≥98.5 % |
Silicate (as SiO2) | ≤50 ppm |
Magnesium carbonate | ≤100 ppm |
Iron (Fe) | ≤5 ppm |
Fines | ≤1.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...