Toluene, extra dry over Molecular Sieve 99.85%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

36441-0010 36441-1000 36441-5000 364410025
TS36441-0010EA 85 USD
TS36441-0010 TS36441-1000 TS36441-5000 TS364410025
Toluene, extra dry over Molecular Sieve 99.85%, AcroSeal™
Toluene
Formula: C₆H₅CH₃
Boiling Pt: 111 °C (760 mmHg)
Melting Pt: –95 °C
Density: 0.866 g/cm³
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.8 %
Acidity (HCl) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR