2,2,4-Trimethylpentane 99% for spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: IsobutyltrimethylmethaneIsooctaneIsobutyltrimethylethane

16791-0025 167915000
TS16791-0025EA 235.48 USD
TS16791-0025 TS167915000
2,2,4-Trimethylpentane 99% for spectroscopy
2,2,4-Trimethylpentane
Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 99.2 °C (1013 hPa)
Melting Pt: –107 °C
Density: 0.69 g/cm³
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/00/81/740081.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.02 %
GC ≥98.5 %
UV at 210 nm A: ≤1
UV at 220 nm A: ≤0.2
UV at 230 nm A: ≤0.1
UV at 240 nm A: ≤0.04
UV at 250 nm A: ≤0.01
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR