n-Pentane 95%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

43403-0250 AC43403-0050
TS43403-0250EA 799 USD
TS43403-0250 TS43403-0050
n-Pentane 95%, Technical Grade
n-Pentane
Formula: H₃C(CH₂)₃CH₃
Boiling Pt: 36 °C (760 mmHg)
Melting Pt: –130 °C
Density: 0.626 g/cm³
Flash Pt: –49 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
UN: 1265
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3570 to 1.3590 (20°C, 589 nm)
GC ≥94 %
UV at 400 nm A: ≤0.009
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR