n-Nonane 99%, pure

Supplier: Thermo Scientific Chemicals
Danger

12911-1000 129110025 129115000
TS12911-1000EA 142 USD
TS12911-1000 TS129110025 TS129115000
n-Nonane 99%, pure
n-Nonane
Formula: H₃C(CH₂)₇CH₃
Boiling Pt: 151 °C (760 mmHg)
Melting Pt: –53 °C
Density: 0.718 g/cm³
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00009574
CAS Number: 111-84-2
EINECS: 203-913-4
UN: 1920
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4060 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR