n-Decane 99+%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
11187-0025
11187-5000
AC11187-1000
TS11187-0025EA
735
USD
TS11187-0025
TS11187-5000
TS11187-1000
n-Decane 99+%, pure
n-Decane
Formula:
H₃C(CH₂)₈CH₃ MW: 142.28 g/mol Boiling Pt: 174 °C (760 mmHg) Melting Pt: –30 °C Density: 0.735 g/cm³ Flash Pt: 46 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008954 CAS Number: 124-18-5 EINECS: 204-686-4 UN: 2247 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4100 to 1.4130 (20°C, 589 nm) |
GC | ≥99.0 % |
Composition of fatty acids | 0.5 to 3.0 % (GC) Lignoceric acid |
Acid value | ≤0.5 mg KOH/g |
Peroxide value | ≤5 meq O2/kg |
Unsaponifiable matter | ≤1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...