Glycerine 99.6% ACS

Supplier: Thermo Scientific Chemicals

41098-0025 41098-0100
TS41098-0025EA 309 USD
TS41098-0025 TS41098-0100 TS410985000
Glycerine 99.6% ACS
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
Boiling Pt: 290 °C (760 mmHg)
Melting Pt: 18 °C
Density: 1.261 g/cm³
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4750 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥99.5 %
Color scale ≤10 APHA
Residue after ignition ≤0.005 %
Substance darkened by H2SO4 Passes test
Sulfate (SO4) ≤10 ppm
Chlorinated cmpds (as Cl) ≤0.003 %
Heavy metals (as Pb) ≤2 ppm
Impurity (Acrolein and glucose) Passes test
Fatty acid esters ≤0.05 % (as Butyric acid)
Neutrality Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR