2-Propanol 99+%, meets analytical specification of Ph.Eur.

Supplier: Thermo Scientific Chemicals
Danger

39698-0025 39698-0100 396980010 396980250 396982500
TS39698-0025EA 89 USD
TS39698-0025 TS39698-0100 TS396980010 TS396980250 TS396982500
2-Propanol 99+%, meets analytical specification of Ph.Eur.
2-Propanol
Formula: (CH₃)₂CHOH
Boiling Pt: 81…83 °C (760 mmHg)
Melting Pt: –89.5 °C
Density: 0.785 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid (Passes test)
Infrared spectrum Conforms
Refractive index 1.376 to 1.379 (20°C, 589 nm)
Water ≤0.5 %
GC ≥99.0 %
UV at 310 nm A: ≤0.01
Relative density (20/20°C) 0.785 to 0.789
Specific gravity (25°C) 0.783 to 0.787
Identification D Passes test
Non-volatile matter ≤20 ppm
Related substances ≤0.3 %
Acidity/alkalinity Passes test
Benzene ≤2 ppm
Peroxides Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR