2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol
42383-0010
42383-0025
42383-0040
42383-0200
42383-0250
42383-5000
TS42383-0010EA
82
USD
TS42383-0010
TS42383-0025
TS42383-0040
TS42383-0200
TS42383-0250
TS42383-5000
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
2-Propanol
Formula:
(CH₃)₂CHOH Boiling Pt: 82 °C (1013 hPa) Melting Pt: –89.5 °C Density: 0.785 g/cm³ Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 UN: 1219 ADR: 3,II |
|
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.2 % (K.F.) |
Boiling point | 81 to 83 °C |
Density | (20/20 °C) 0.784 to 0.787 |
Solubility | (in water) Passes test |
GC | ≥99.5 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Carbonyl compounds | ≤0.002 % (acetone) |
Titrable acid or base | ≤0.0001 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...