Trifluoroacetic acid 99+% for HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TFA

43229-1000 43229-5000
TS43229-1000EA 442 USD
TS43229-1000 TS43229-5000
Trifluoroacetic acid 99+% for HPLC
Trifluoroacetic acid
Formula: C₂HF₃O₂
MW: 114.02 g/mol
Boiling Pt: 71.8
Melting Pt: -15
Density: 1.535 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
UN: 2699
ADR: 8 I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.05 %
UV at 320 nm A: ≤0.03
Color scale ≤10 APHA
Residue after evaporation ≤5 ppm
Titration with NaOH ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR