Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal™

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methane dichloride, Methylene chloride, Methylene chloride

34846-0010 34846-5000 348460025 348461000
TS34846-0010EA 91 USD
TS34846-0010 TS34846-5000 TS348460025 TS348461000
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal™
Dichloromethane
Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8
Melting Pt: -95
Density: 1.325 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.8 %
Stabilizer Approx. 50 ppm Amylene


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR