Citric acid, anhydrous 99% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid
, Hydroxytricarballylic acid
42356-5000
42356-0100
42356-0020
TS42356-0020EA
188
USD
TS42356-0020
TS42356-0100
TS42356-5000
Citric acid, anhydrous 99% ACS
Citric acid
Formula:
HOC(COOH)(CH₂COOH)₂ MW: 192.13 g/mol Boiling Pt: 199.6 °C (1013 hPa) Melting Pt: –12 °C Density: 1.08 g/cm³ Flash Pt: 77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011669 CAS Number: 77-92-9 EINECS: 202-259-7 REACH: 01-2119457026-42-0000 Merck Index: 13,06052 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5160 to 1.5180 (20°C, 589 nm) |
GC | ≥98.5 % |
Specific gravity | (25/25°C) 1.084 to 1.088 |
Acid value | ≤1 mg KOH/g |
UV | at 300 nm A: ≤0.01 |
UV | 1 cm cell vs water |
UV | at 245 nm A: ≤1.00 |
Color scale | ≤10 APHA |
Stabilizer | 0.5 to 1.0 % Ethanol |
Residue after evaporation | ≤0.0002 % |
Free chlorine | ≤0.00003 % (as Cl) |
Free acid (HCl) | ≤0.001 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...