Ammonium acetate 98+% for HPLC

Supplier: Thermo Scientific Chemicals

21999-0500 21999-2500
TS21999-0500EA 79 USD
TS21999-0500 TS21999-2500
Ammonium acetate 98+% for HPLC
Formula: H₃CCOONH₄
Melting Pt: 114 °C
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR