Stearic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Octadecanoic acid, Octadecylic acid
17449-0025
17449-0010
17449-0100
TS17449-0025EA
103
USD
TS17449-0025
TS17449-0010
TS17449-0100
Stearic acid 99%
Stearic acid
Formula:
C₁₈H₃₆O₂ Boiling Pt: 269…270 °C (760 mmHg) Melting Pt: 67…69 °C Density: 0.96 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002752 CAS Number: 57-11-4 EINECS: 214-738-8 UN: 3334 ADR: 9,III Merck Index: 13,08882 |
|
Specification Test Results
Appearance | Colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5030 to 1.5050 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...