Formic acid 88% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methanoic acidAminic Acid

410775000 41077-0025
TS410775000EA 98.8 USD
TS410775000 TS41077-0025
Formic acid 88% ACS
Formic acid
Formula: HCO₂H
MW: 46.03 g/mol
Boiling Pt: 101 °C (760 mmHg)
Melting Pt: 8 °C
Density: 1.22 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/37/23/723723.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3694 to 1.3714 (20°C, 589 nm)
Color scale ≤15 APHA
Residue after evaporation ≤0.002 %
Titration with NaOH ≥88.0 %
Dilution test passes test
Chloride (Cl) ≤10 ppm
Sulfite (SO3) passes test
Sulfate (SO4) ≤20 ppm
Ammonium (NH4) ≤50 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Free acid (CH3COOH) ≤0.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR