Isopropyl acetate 99+% for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acetic acid isopropyl ester
38950-0010
389500025
TS38950-0010EA
121
USD
TS38950-0010
TS389500025
Isopropyl acetate 99+% for analysis
Isopropyl acetate
Formula:
C₅H₁₀O₂ Boiling Pt: 88.8 °C (760 mmHg) Melting Pt: –73 °C Density: 0.872 g/cm³ Flash Pt: 4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008877 CAS Number: 108-21-4 EINECS: 203-561-1 UN: 1220 ADR: 3,II Merck Index: 13,05225 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3760 to 1.3780 (20°C, 589 nm) |
Water | ≤0.1 % (Coulometric) |
GC | ≤0.1 % 2-Propanol |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.002 % |
Cadmium (Cd) | ≤0.05 ppm |
Calcium (Ca) | ≤0.5 ppm |
Chromium (Cr) | ≤0.02 ppm |
Cobalt (Co) | ≤0.02 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.1 ppm |
Magnesium (Mg) | ≤0.1 ppm |
Manganese (Mn) | ≤0.02 ppm |
Nickel (Ni) | ≤0.02 ppm |
Zinc (Zn) | ≤0.1 ppm |
Acidity (CH3COOH) | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...