Acetonitrile 99.9%, meets analytical specification of Ph.Eur. for HPLC, far-UV
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
26826-0025
268260010
TS26826-0025EA
275
USD
TS26826-0025
TS268260010
Acetonitrile 99.9%, meets analytical specification of Ph.Eur. for HPLC, far-UV
Acetonitrile
Formula:
H₃CCN Boiling Pt: 81.6 °C (1013 hPa) Melting Pt: –46 °C Density: 0.781 g/cm³ Flash Pt: 2 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.02 % |
GC | ≥99.9 % |
UV | 1 cm cell vs HPLC-grade water |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.0002 % |
Acidity/alkalinity | ≤0.0008 meq/g |
Filtered to 0.2 micrometer | Filtered |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...